Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Upf3b
Entrez Gene ID
313449
Gene Name
UPF3 regulator of nonsense transcripts homolog B (yeast)
Gene Aliases
RGD1560264
Chromosome Location
Chr.X: 123713327 - 123731426 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257455.2 XP_006257517.1 8 - 9 982 102
XM_008773416.1 XP_008771638.1 8 - 9 982 102

View Full Product Details