Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lonrf3
Entrez Gene ID
298322
Gene Name
LON peptidase N-terminal domain and ring finger 3
Gene Aliases
RGD1565451
Chromosome Location
Chr.X: 122937987 - 122971578 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191585.1 NP_001178514.1 5 - 6 1546 116
XM_006257447.2 XP_006257509.1 6 - 7 1750 116

View Full Product Details