Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rasip1
Entrez Gene ID
292912
Gene Name
Ras interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 101608995 - 101621648 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106261.2 NP_001099731.1 5 - 6 1240 136
XM_006229030.2 XP_006229092.1 7 - 8 2325 136
GenBank mRNA BC168876.1 - 5 - 6 1240 136

View Full Product Details