Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lekr1
Entrez Gene ID
361953
Gene Name
leucine, glutamate and lysine rich 1
Gene Aliases
RGD1562738, RGD1565425
Chromosome Location
Chr.2: 157452004 - 157621908 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115029.2 NP_001108501.2 2 - 3 507 134
XM_006232473.1 XP_006232535.1 2 - 3 169 134
GenBank mRNA BC107462.1 - 2 - 3 471 134
BC158627.1 - 2 - 3 507 134

View Full Product Details