Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fhdc1
Entrez Gene ID
295161
Gene Name
FH2 domain containing 1
Gene Aliases
RGD1311955
Chromosome Location
Chr.2: 183450608 - 183487820 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106437.1 NP_001099907.1 6 - 7 1117 79

View Full Product Details