The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079711.1 | NP_001073179.1 | 8 - 9 | 1062 | 66 |
XM_006237502.2 | XP_006237564.1 | 8 - 9 | 1045 | 66 | |
GenBank mRNA | BC128747.1 | - | 8 - 9 | 1062 | 66 |