The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013927.1 | NP_001013949.1 | 3 - 4 | 598 | 109 |
GenBank mRNA | BC085851.1 | - | 3 - 4 | 598 | 109 |