Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100360076
Entrez Gene ID
100360076
Gene Name
putative taste receptor T2R14-like
Gene Aliases
-
Chromosome Location
Chr.4: 167051512 - 167052477 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729433.1 XP_002729479.1 1 - 1 751 137

View Full Product Details