The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199207.1 | NP_954677.1 | 30 - 31 | 3933 | 136 |
XM_006237147.2 | XP_006237209.1 | 29 - 30 | 3894 | 136 | |
XM_006237149.2 | XP_006237211.1 | 29 - 30 | 3855 | 136 | |
GenBank mRNA | AY257251.1 | - | 30 - 31 | 3933 | 136 |