Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam21c
Entrez Gene ID
297530
Gene Name
family with sequence similarity 21, member C
Gene Aliases
Fam21, NP61201
Chromosome Location
Chr.4: 148139512 - 148197361 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199207.1 NP_954677.1 8 - 9 810 198
XM_006237147.2 XP_006237209.1 8 - 9 834 198
XM_006237149.2 XP_006237211.1 8 - 9 834 198
GenBank mRNA AY257251.1 - 8 - 9 810 198

View Full Product Details