Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Emg1
Entrez Gene ID
312706
Gene Name
EMG1 N1-specific pseudouridine methyltransferase
Gene Aliases
Grcc2f
Chromosome Location
Chr.4: 157222366 - 157230647 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107888.2 NP_001101358.2 1 - 2 304 82

View Full Product Details