The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031515.3 | NP_113703.1 | 3 - 4 | 448 | 88 |
XM_008763354.1 | XP_008761576.1 | 4 - 5 | 630 | 88 | |
GenBank mRNA | BC126086.1 | - | 4 - 5 | 586 | 88 |
FQ230946.1 | - | 4 - 5 | 629 | 88 | |
U09793.1 | - | 3 - 4 | 448 | 88 |