The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053018.1 | NP_444177.1 | 7 - 8 | 671 | 70 |
GenBank mRNA | BC161804.1 | - | 7 - 8 | 646 | 70 |
BG670145.1 | - | 3 - 4 | 263 | 70 | |
CF111078.1 | - | 7 - 8 | 680 | 70 | |
FQ212497.1 | - | 7 - 8 | 695 | 70 | |
FQ213039.1 | - | 7 - 8 | 720 | 70 | |
FQ213302.1 | - | 7 - 8 | 689 | 70 | |
FQ215636.1 | - | 7 - 8 | 683 | 70 | |
FQ219799.1 | - | 7 - 8 | 694 | 70 | |
FQ229965.1 | - | 7 - 8 | 742 | 70 | |
X76489.1 | - | 7 - 8 | 610 | 70 |