Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec9a
Entrez Gene ID
502901
Gene Name
C-type lectin domain family 9, member A
Gene Aliases
RGD1562513
Chromosome Location
Chr.4: 163173597 - 163189532 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109354.1 NP_001102824.1 1 - 2 185 66
XM_006237099.2 XP_006237161.1 1 - 2 1061 66
XM_008763326.1 XP_008761548.1 1 - 2 62 66
GenBank mRNA GU357486.1 - 1 - 2 89 66

View Full Product Details