Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcf7
Entrez Gene ID
363595
Gene Name
transcription factor 7 (T-cell specific, HMG-box)
Gene Aliases
-
Chromosome Location
Chr.10: 37616022 - 37647851 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006246362.2 XP_006246424.1 3 - 4 671 137
XM_006246363.2 XP_006246425.1 3 - 4 681 137
XM_006246364.2 XP_006246426.1 3 - 4 675 137
XM_006246365.2 XP_006246427.1 3 - 4 671 137
XM_008767741.1 XP_008765963.1 3 - 4 2495 137

View Full Product Details