Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnmtl1
Entrez Gene ID
360569
Gene Name
RNA methyltransferase like 1
Gene Aliases
RGD1309077
Chromosome Location
Chr.10: 64398339 - 64405017 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108282.1 NP_001101752.1 2 - 3 566 71

View Full Product Details