Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam57a
Entrez Gene ID
100360533
Gene Name
family with sequence similarity 57, member A
Gene Aliases
RGD1307493
Chromosome Location
Chr.10: 64358828 - 64367679 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002727796.4 XP_002727842.2 4 - 5 2144 77

View Full Product Details