Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nup85
Entrez Gene ID
287830
Gene Name
nucleoporin 85
Gene Aliases
Pcnt1
Chromosome Location
Chr.10: 104118999 - 104137258 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025401.1 NP_001020572.1 8 - 9 759 69
GenBank mRNA BC098031.1 - 8 - 9 759 69

View Full Product Details