Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Etv4
Entrez Gene ID
360635
Gene Name
ets variant 4
Gene Aliases
Pea3
Chromosome Location
Chr.10: 89685058 - 89700314 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108299.1 NP_001101769.1 12 - 13 1504 83
XM_006247369.2 XP_006247431.1 12 - 13 1577 83
XM_006247370.2 XP_006247432.1 12 - 13 1525 83
XM_006247371.2 XP_006247433.1 12 - 13 1485 83
XM_006247372.2 XP_006247434.1 12 - 13 1332 83
XM_008768085.1 XP_008766307.1 12 - 13 1574 83
XM_008768086.1 XP_008766308.1 12 - 13 1560 83
GenBank mRNA BC168950.1 - 12 - 13 1378 83

View Full Product Details