The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022393.2 | NP_071788.2 | 8 - 9 | 958 | 76 |
XM_006246803.2 | XP_006246865.1 | 8 - 9 | 996 | 76 | |
XM_008767867.1 | XP_008766089.1 | 8 - 9 | 996 | 76 | |
XM_008767868.1 | XP_008766090.1 | 8 - 9 | 840 | 76 | |
XM_008767869.1 | XP_008766091.1 | 8 - 9 | 837 | 76 | |
GenBank mRNA | BC091151.1 | - | 8 - 9 | 958 | 76 |
FQ213042.1 | - | 8 - 9 | 830 | 76 | |
FQ221204.1 | - | 8 - 9 | 847 | 76 | |
FQ228437.1 | - | 8 - 9 | 816 | 76 | |
J05495.1 | - | 8 - 9 | 76 |