Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cd7
Entrez Gene ID
303747
Gene Name
Cd7 molecule
Gene Aliases
-
Chromosome Location
Chr.10: 110229931 - 110232812 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107074.1 NP_001100544.1 3 - 4 581 57
GenBank mRNA BC167030.1 - 3 - 4 553 57
FQ226313.1 - 3 - 4 583 57
FQ226643.1 - 3 - 4 583 57
FQ227263.1 - 3 - 4 583 57

View Full Product Details