Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wscd1
Entrez Gene ID
287466
Gene Name
WSC domain containing 1
Gene Aliases
RGD1308212
Chromosome Location
Chr.10: 58342393 - 58371199 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024234.1 NP_001019405.1 5 - 6 1403 76
GenBank mRNA BC094520.1 - 5 - 6 1403 76

View Full Product Details