Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pitpna
Entrez Gene ID
29525
Gene Name
phosphatidylinositol transfer protein, alpha
Gene Aliases
Pitpn
Chromosome Location
Chr.10: 63731767 - 63772049 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017231.1 NP_058927.1 10 - 11 957 94
GenBank mRNA BC070945.1 - 10 - 11 985 94
M25758.1 - 10 - 11 957 94

View Full Product Details