Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rundc3a
Entrez Gene ID
303569
Gene Name
RUN domain containing 3A
Gene Aliases
GCRP, Rap2ip
Chromosome Location
Chr.10: 90342029 - 90351172 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198758.2 NP_942053.2 2 - 3 458 85
XM_006247335.2 XP_006247397.1 2 - 3 480 85
XM_006247337.2 XP_006247399.1 2 - 3 480 85
XM_006247339.2 XP_006247401.1 2 - 3 480 85
XM_008768059.1 XP_008766281.1 2 - 3 480 85
GenBank mRNA CB797318.1 - 1 - 2 139 85
DY319760.1 - 2 - 3 492 85

View Full Product Details