Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eme2
Entrez Gene ID
302982
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Gene Aliases
RGD1304924
Chromosome Location
Chr.10: 14253552 - 14256993 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750789.3 XP_003750837.1 6 - 7 828 129
XM_008767602.1 XP_008765824.1 6 - 7 828 129
XM_008767605.1 XP_008765827.1 6 - 7 794 129
GenBank mRNA BC158841.1 - 5 - 6 925 129

View Full Product Details