Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmb6
Entrez Gene ID
29666
Gene Name
proteasome (prosome, macropain) subunit, beta type 6
Gene Aliases
Psmb6l
Chromosome Location
Chr.10: 57131347 - 57133685 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057099.3 NP_476440.2 2 - 3 181 100
XM_008767811.1 XP_008766033.1 2 - 3 212 100
GenBank mRNA BC058451.1 - 2 - 3 185 100
D10754.1 - 2 - 3 170 100
FQ209811.1 - 2 - 3 184 100
FQ217385.1 - 2 - 3 184 100
FQ224639.1 - 2 - 3 184 100

Gene Symbol
LOC100360846
Entrez Gene ID
100360846
Gene Name
proteasome subunit beta type 6-like
Gene Aliases
-
Chromosome Location
Chr.2: 96031948 - 96033565 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729028.3 XP_002729074.2 2 - 2 329 100

View Full Product Details