Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd40
Entrez Gene ID
690586
Gene Name
ankyrin repeat domain 40
Gene Aliases
-
Chromosome Location
Chr.10: 82032656 - 82045937 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134699.1 NP_001128171.1 1 - 2 332 64
GenBank mRNA BC166790.1 - 1 - 2 332 64

View Full Product Details