The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134699.1 | NP_001128171.1 | 4 - 5 | 1142 | 92 |
GenBank mRNA | BC166790.1 | - | 4 - 5 | 1142 | 92 |