Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc8
Entrez Gene ID
303376
Gene Name
fibronectin type III domain containing 8
Gene Aliases
RGD1310708
Chromosome Location
Chr.10: 70242852 - 70250990 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107030.1 NP_001100500.1 1 - 2 280 114

View Full Product Details