Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Utp18
Entrez Gene ID
303456
Gene Name
UTP18 small subunit (SSU) processome component homolog (yeast)
Gene Aliases
RGD1309957, Wdr50
Chromosome Location
Chr.10: 81560929 - 81588151 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135039.1 NP_001128511.1 13 - 14 1703 101
XM_008768044.1 XP_008766266.1 13 - 14 1959 102
GenBank mRNA CK843896.1 - 1 - 2 101

View Full Product Details