Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sgca
Entrez Gene ID
303468
Gene Name
sarcoglycan, alpha (dystrophin-associated glycoprotein)
Gene Aliases
-
Chromosome Location
Chr.10: 82770905 - 82785142 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107039.1 NP_001100509.1 4 - 5 556 67
XM_006247191.2 XP_006247253.1 3 - 4 380 67
XM_006247192.1 XP_006247254.1 3 - 4 284 67
GenBank mRNA FQ215532.1 - 3 - 4 342 67

View Full Product Details