Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eno3
Entrez Gene ID
25438
Gene Name
enolase 3, beta, muscle
Gene Aliases
BBE, MSE
Chromosome Location
Chr.10: 57278268 - 57283661 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012949.2 NP_037081.2 11 - 12 1287 90
XM_006246599.2 XP_006246661.1 11 - 12 1358 90
XM_006246600.2 XP_006246662.1 11 - 12 1338 90
GenBank mRNA BC083566.1 - 11 - 12 1274 90
FQ214810.1 - 10 - 11 1266 90
FQ214945.1 - 11 - 12 1305 90
FQ214984.1 - 11 - 12 1299 90
FQ215034.1 - 7 - 8 965 90
FQ215197.1 - 7 - 8 964 90
FQ215221.1 - 10 - 11 1199 90
FQ215310.1 - 7 - 8 965 90
FQ215357.1 - 11 - 12 1310 90
FQ215526.1 - 11 - 12 1298 90
FQ215607.1 - 7 - 8 964 90
FQ215648.1 - 11 - 12 1225 90
FQ215780.1 - 7 - 8 965 90
FQ215843.1 - 10 - 11 1265 90
FQ215865.1 - 11 - 12 1297 90
FQ216020.1 - 7 - 8 964 90
FQ216150.1 - 10 - 11 1266 90
FQ216382.1 - 11 - 12 1300 90
FQ216660.1 - 10 - 11 1265 90
FQ216781.1 - 7 - 8 964 90
FQ217095.1 - 11 - 12 1297 90
FQ223831.1 - 7 - 8 964 90
FQ224938.1 - 11 - 12 1297 90
Y00979.1 - 10 - 11 1249 90

View Full Product Details