Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gfer
- Entrez Gene ID
-
27100
- Gene Name
- growth factor, augmenter of liver regeneration
- Gene Aliases
-
ALR
- Chromosome Location
-
Chr.10: 14059347 - 14061703 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_013222.2 |
NP_037354.2 |
1 - 2 |
319 |
75 |
GenBank mRNA |
D30735.1 |
- |
1 - 2 |
319 |
75 |
- Gene Symbol
- LOC100912596
- Entrez Gene ID
-
100912596
- Gene Name
- FAD-linked sulfhydryl oxidase ALR-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10|NW_007906257: 10 - 2122 on Build Rnor_6.0
View Full Product Details