Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgcd
Entrez Gene ID
497892
Gene Name
sarcoglycan, delta (dystrophin-associated glycoprotein)
Gene Aliases
-
Chromosome Location
Chr.10: 32062946 - 32471454 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134826.1 NP_001128298.1 3 - 4 565 100

View Full Product Details