Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsr1
Entrez Gene ID
100360406
Gene Name
TSR1, 20S rRNA accumulation, homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.10: 61746070 - 61757518 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002727774.2 XP_002727820.1 7 - 8 1337 112

View Full Product Details