Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tlcd2
Entrez Gene ID
497955
Gene Name
TLC domain containing 2
Gene Aliases
RGD1562631
Chromosome Location
Chr.10: 62303124 - 62306198 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006246868.2 XP_006246930.1 2 - 3 907 139
XM_006246869.2 XP_006246931.1 2 - 3 907 139
XM_006246870.2 XP_006246932.1 2 - 3 907 139
XM_573142.6 XP_573142.4 2 - 3 907 139
GenBank mRNA FQ219135.1 - 2 - 3 379 139

View Full Product Details