Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a10
Entrez Gene ID
170943
Gene Name
solute carrier family 25 (mitochondrial carrier; dicarboxylate transporter), member 10
Gene Aliases
Dic
Chromosome Location
Chr.10: 109665682 - 109673143 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133418.1 NP_596909.1 9 - 10 731 63
GenBank mRNA AJ223355.1 - 9 - 10 731 63
BC081734.1 - 9 - 10 749 63

View Full Product Details