Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc13a5
Entrez Gene ID
266998
Gene Name
solute carrier family 13 (sodium-dependent citrate transporter), member 5
Gene Aliases
Nact
Chromosome Location
Chr.10: 58810054 - 58835475 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170668.1 NP_733768.1 9 - 10 1303 165
XM_006246602.2 XP_006246664.1 10 - 11 2529 165
GenBank mRNA AF522186.1 - 9 - 10 1303 165

View Full Product Details