The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111064.1 | NP_001104534.1 | 8 - 9 | 1783 | 71 |
GenBank mRNA | AI602815.1 | - | 4 - 5 | 318 | 71 |