Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dach1
Entrez Gene ID
306096
Gene Name
dachshund family transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.15: 82138401 - 82285518 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770929.1 XP_008769151.1 2 - 3 309 69

View Full Product Details