The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107281.1 | NP_001100751.1 | 7 - 8 | 1238 | 124 |
XM_006252408.2 | XP_006252470.1 | 7 - 8 | 1189 | 124 | |
XM_006252409.2 | XP_006252471.1 | 7 - 8 | 1184 | 124 | |
XM_006252410.2 | XP_006252472.1 | 7 - 8 | 1255 | 124 | |
XM_006252413.2 | XP_006252475.1 | 6 - 7 | 1037 | 124 | |
XM_006252415.2 | XP_006252477.1 | 6 - 7 | 1034 | 124 | |
XM_008770870.1 | XP_008769092.1 | 6 - 7 | 2991 | 124 | |
XM_008770871.1 | XP_008769093.1 | 8 - 9 | 1201 | 124 |