Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajc3
Entrez Gene ID
63880
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 3
Gene Aliases
-
Chromosome Location
Chr.15: 104186918 - 104226236 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022232.1 NP_071568.1 4 - 5 394 69
GenBank mRNA AB017702.3 - 4 - 5 394 69
BC061764.1 - 4 - 5 489 69

View Full Product Details