Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx11
Entrez Gene ID
303493
Gene Name
sorting nexin 11
Gene Aliases
-
Chromosome Location
Chr.10: 84628420 - 84639126 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012012.2 NP_001012012.2 2 - 3 141 66
XM_006247195.2 XP_006247257.1 3 - 4 534 66
XM_008768047.1 XP_008766269.1 3 - 4 885 66
GenBank mRNA BC086543.1 - 2 - 3 141 66
FQ212083.1 - 2 - 3 112 66

View Full Product Details