Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spag5
Entrez Gene ID
252918
Gene Name
sperm associated antigen 5
Gene Aliases
Deepest
Chromosome Location
Chr.10: 65566228 - 65585773 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044224.1 NP_001037689.1 14 - 15 2579 88
GenBank mRNA AB231661.1 - 14 - 15 2579 88
AF111111.1 - 2 - 3 151 88
BC166408.1 - 14 - 15 2569 88

View Full Product Details