Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tlk2
Entrez Gene ID
303592
Gene Name
tousled-like kinase 2
Gene Aliases
-
Chromosome Location
Chr.10: 93412584 - 93507277 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191652.1 NP_001178581.1 17 - 18 2072 108
XM_006247574.2 XP_006247636.1 17 - 18 3415 108
XM_006247576.2 XP_006247638.1 17 - 18 1968 108
XM_008768348.1 XP_008766570.1 16 - 17 1933 108
XM_008768349.1 XP_008766571.1 15 - 16 1841 108
XM_008768350.1 XP_008766572.1 12 - 13 1265 108
GenBank mRNA BC086544.1 - 7 - 8 677 108

View Full Product Details