The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191652.1 | NP_001178581.1 | 20 - 21 | 2499 | 72 |
XM_006247574.2 | XP_006247636.1 | 20 - 21 | 3842 | 72 | |
XM_006247576.2 | XP_006247638.1 | 20 - 21 | 2395 | 72 | |
XM_008768348.1 | XP_008766570.1 | 19 - 20 | 2360 | 72 | |
XM_008768349.1 | XP_008766571.1 | 18 - 19 | 2268 | 72 | |
XM_008768350.1 | XP_008766572.1 | 15 - 16 | 1692 | 72 | |
GenBank mRNA | BC086544.1 | - | 10 - 11 | 1104 | 72 |