The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173128.1 | NP_775151.1 | 6 - 7 | 1204 | 101 |
GenBank mRNA | M63665.1 | - | 6 - 7 | 1204 | 101 |