Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ovca2
Entrez Gene ID
497954
Gene Name
ovarian tumor suppressor candidate 2
Gene Aliases
RGD1564623
Chromosome Location
Chr.10: 62017347 - 62020509 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109036.1 NP_001102506.1 1 - 2 194 120

View Full Product Details