Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Inpp5k
Entrez Gene ID
287533
Gene Name
inositol polyphosphate-5-phosphatase K
Gene Aliases
Skip
Chromosome Location
Chr.10: 63776268 - 63795902 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013859.1 NP_001013881.1 7 - 8 891 99
GenBank mRNA BC083572.1 - 7 - 8 891 99
FQ228522.1 - 5 - 6 1800 99

View Full Product Details