Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tex2
Entrez Gene ID
303611
Gene Name
testis expressed 2
Gene Aliases
-
Chromosome Location
Chr.10: 94697970 - 94807539 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006247645.1 XP_006247707.1 5 - 6 2567 146
XM_006247646.2 XP_006247708.1 5 - 6 3399 146
XM_006247647.1 XP_006247709.1 5 - 6 2567 146
GenBank mRNA BC083806.1 - 4 - 5 951 146
BC097967.1 - 1 - 2 229 146

View Full Product Details